Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Myeloperoxidase Protein

Catalog No. 3667MP250
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3667MP250 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 3667MP250 Supplier R&D Systems™ Supplier No. 3667MP250
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Measured by its ability to oxidize guaiacol in the presence of hydrogen peroxide. Capeillere-Blandin, C. (1998) Biochem J. 336 :395. The specific activity is >8,000 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number AAR99349
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 4353
Molecular Weight (g/mol) M.W. (Observed): 60-70 kDa and 85-95 kDa, reducing conditions; M.W. (theoretical): 81 kDa
Name Myeloperoxidase/MPO
Quantity 250 μg
Source Mouse myeloma cell line, NS0-derived mouse Myeloperoxidase/MPO protein Met16-Thr718, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Mpo
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in sterile, deionized water.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Myeloperoxidase/MPO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.