Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Nectin-2/CD112 Protein
SDP

Catalog No. 3869N2050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3869N2050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 3869N2050 Supplier R&D Systems™ Supplier No. 3869N2050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Mouse Nectin-2/CD112 is coated at 1 μg/mL (100 μL/well), the concentration of Recombinant Mouse DNAM-1 Fc Chimera (Catalog # 4436-DN ) that produces 50% optimal binding response is 0.3-1.8 μg/mL.

Specifications

Accession Number P32507
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5819
Molecular Weight (g/mol) M.W. (Observed): 45-50 kDa, reducing conditions; M.W. (theoretical): 35.4 kDa
Name Nectin-2/CD112
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse Nectin-2/CD112 protein Gln32-Gly351, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Monomer
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol Nectin2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Protein Nectin-2/CD112
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.