Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse NELL2 Protein
SDP

Catalog No. 9000NL050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-00N-L050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-00N-L050 Supplier R&D Systems™ Supplier No. 9000NL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse NELL2 Protein is derived from NS0. The Recombinant Mouse NELL2 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q61220
For Use With (Application) Bioactivity
Gene ID (Entrez) 4753
Molecular Weight (g/mol) Observed M.W.: 105-130kDa, reducing conditions, Theoretical M.W.: 90kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse NELL2 protein Leu25-Leu819,with an N-Terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Mel91, nel (chicken)-like 2, NEL-like 2 (chicken), NEL-like protein 2, Nel-related protein 2, NRP2, NRP2neural epidermal growth factor-like 2, protein kinase C-binding protein NELL2
Common Name NELL2
Gene Symbol Nell2
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of C3H10T1/2 mouse embryonic fibroblast cells. The ED50 for this effect is typically 0.3-1.5μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Leu25-Leu819, with an N-Terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.