Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Netrin-1 Protein

Catalog No. 1109N1025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS and EDTA with Trehalose.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
1109N1025 Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
1109N1025CF Lyophilized from a 0.2μm filtered solution in PBS and EDTA with Trehalose.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 1109N1025 Supplier R&D Systems™ Supplier No. 1109N1025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Mouse Netrin-1 Protein is derived from NS0. The Recombinant Mouse Netrin-1 Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS and EDTA with BSA as a carrier protein.
Gene ID (Entrez) 18208
Molecular Weight (g/mol) 68.2 kDa
Name Netrin-1
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse Netrin-1 protein, (N-terminus) Mouse Netrin-1 (Val22-Ala603) Accession &Num; AAC52971 IEGR GGGSGGGSGGGS 10-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.