Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Neuregulin-1/NRG1 Protein

Catalog No. 9875NR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
98-75N-R050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 98-75N-R050 Supplier R&D Systems™ Supplier No. 9875NR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

  • Derived from NS0
  • Validated for the following applications: Bioactivity.

Specifications

Accession Number XP_006509143
For Use With (Application) Bioactivity
Gene ID (Entrez) 3084
Molecular Weight (g/mol) 42 to 49 kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse Neuregulin-1/NRG1 protein Ser20-Lys246, with a C-terminal 6-His tag
Storage Requirements 12 months from date of receipt, -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution 3 months, -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Purity or Quality Grade > 95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.