Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse NKG2A (CHO-expressed) Fc Protein
SDP

Catalog No. 9198NK050 Shop All R&D Systems Products
Change view
Click to view available options
Recombinant Mouse NKG2A Fc Chimera Protein, Carrier-Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
91-98N-K050 Recombinant Mouse NKG2A Fc Chimera Protein, Carrier-Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 91-98N-K050 Supplier R&D Systems™ Supplier No. 9198NK050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse NKG2A Fc Chimera Protein is derived from CHO. The Recombinant Mouse NKG2A Fc Chimera Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

Specifications

For Use With (Application) Bioactivity
Source Chinese Hamster Ovary cell line,CHO-derived mouse NKG2A/CD159a protein, (N-terminus) MDP Mouse IgG2A (Glu98-Lys330) IEGR Mouse NKG2A (Ala94-IIe244) Accession &Num; Q9Z202 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.