Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Noggin Protein

Catalog No. 1967NG100CF
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-67N-G100CF 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-67N-G100CF Supplier R&D Systems™ Supplier No. 1967NG100/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

∼2 fold better activity; Lower endotoxin specification; Enhanced purity.

  • Measured by its ability to inhibit BMP-4-induced alkaline phosphatase production by ATDC5 mouse chondrogenic cells.
  • The ED50 for this effect is 0.06 to 0.3 μg/mL in the presence of 50 ng/mL of Recombinant Human BMP-4.

Specifications

Accession Number P97466
For Use With (Application) Bioactivity
Gene ID (Entrez) 9241
Molecular Weight (g/mol) 26 kDa (monomer)
Name NOG, noggin, SYM1, symphalangism 1 (proximal), synostoses (multiple) syndrome 1, SYNS1, SYNS1A
Quantity 100 μg
Source

Mouse myeloma cell line, NS0-derived mouse Noggin protein

Mouse Noggin (Leu20-Cys232) Accession # P97466IEGRGGGSGGGSGGGS10-His tagN-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Gene Symbol Nog
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.