Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse NPRC/NPR3 Fc Chimera Protein
SDP

Catalog No. 10187NR050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-187-NR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-187-NR050 Supplier R&D Systems™ Supplier No. 10187NR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Atrial Natriuretic Peptide Receptor-3 (NPR3), also known as NPR-C or ANPR-C, is one of the three natriuretic peptide receptors

  • Measured by its binding ability in a functional ELISA.
  • When Recombinant Mouse NPRC/NPR3 Fc Chimera is immobilized at 2 μg/mL (100 μL/well), the concentration of Recombinant Mouse Osteocrin (Catalog # 9700-ON) that produces 50% of the optimal binding response is 0.6-3.6 ng/mL.

Specifications

Accession Number P70180
For Use With (Application) Bioactivity
Gene ID (Entrez) 4883
Molecular Weight (g/mol) 78 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived mouse NPRC/NPR3 protein

Mouse NPRC/NPR3 (Glu41-Ala478) Accession # P70180IEGRMDPMouse IgG2a (Glu98-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.