Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse OCAM/NCAM2 His-tag Protein

Catalog No. 10154CM050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-154-CM050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-154-CM050 Supplier R&D Systems™ Supplier No. 10154CM050
Add to Cart
Add to Cart
This item is not returnable. View return policy

OCAM (olfactory cell adhesion molecule), also known as NCAM2 (neural cell adhesion molecule 2) and RNCAM (Rb-8 neural cell adhesion molecule) is an approximately 100 kDa type I transmembrane glycoprotein belonging to the NCAM family within the immunoglobulin superfamily

  • Recombinant Protein
  • Carrier Free (CF)
  • The carrier free version does not contain Bovine Serum Albumin (BSA)

Specifications

Accession Number O35136
For Use With (Application) Bioactivity
Gene ID (Entrez) 4685
Molecular Weight (g/mol) 76.8 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse OCAM/NCAM2 protein Met1-Asn697, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.