Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse PD-1 Fc Chimera Protein

Catalog No. 1021PD100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1021PD100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1021PD100 Supplier R&D Systems™ Supplier No. 1021PD100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. Immobilized Recombinant Mouse PD-1 Fc Chimera at 1 μg/mL (100 μL/well) can bind Recombinant Mouse B7-H1/PD-L1 Fc Chimera (Catalog # 1019-B7 ) with an ED50 of 0.16-0.8 μg/mL.

Specifications

Accession Number Q02242
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCL, NaCl and EDTA with Trehalose.
Gene ID (Entrez) 5133
Molecular Weight (g/mol) M.W. (Observed): 66 kDa, reducing conditions; M.W. (theoretical): 43 kDa (monomer)
Name PD-1 Fc Chimera
Quantity 100 μg
Source Mouse myeloma cell line, NS0-derived mouse PD-1 protein Mouse PD-1 (Leu25-Gln167) (N-terminus) Accession # Q02242 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2
Gene Symbol Pdcd1
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 0.5 mg/mL in sterile water.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PD-1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.