Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Pentraxin 3/TSG-14 Protein

Catalog No. 2166TS025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 .
Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
2166TS025 Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein.
2166TS025CF Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 .
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 2166TS025 Supplier R&D Systems™ Supplier No. 2166TS025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Mouse Pentraxin 3/TSG-14 Protein is derived from NS0. The Recombinant Mouse Pentraxin 3/TSG-14 Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, NaCl and CaCl 2 with BSA as a carrier protein.
Gene ID (Entrez) 19288
Molecular Weight (g/mol) 41 kDa (monomer)
Name Pentraxin 3/TSG-14
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse Pentraxin 3/TSG-14 protein Glu18-Ser381,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Multimer consisting of as many as ten non-covalently and covalently linked subunits
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.