Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse PGLYRP1/PGRP-S His-tag Protein, CF

Catalog No. 2696PGB050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-96P-GB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-96P-GB050 Supplier R&D Systems™ Supplier No. 2696PGB050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Carrier Free Recombinant Protein

Measured by its binding ability in a functional ELISA. When Peptidoglycan is immobilized at 5μg/mL (100μL/well), Recombinant Mouse PGLYRP1/PGRP-S His-tag (Catalog # 2696-PGB) binds with an ED50 of 0.400-4.80 ng/mL.

Specifications

Accession Number O88593.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in MOPS and NaCl with Trehalose.
Gene ID (Entrez) 8993
Molecular Weight (g/mol) M.W. (Observed): 18-21 kDa, under reducing conditions., M.W. (Theoretical): 20 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse PGLYRP1/PGRP-S protein Cys17-Glu182, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias peptidoglycan recognition protein 1, Peptidoglycan recognition protein short, PGLYRP, PGRPpeptidoglycan recognition protein, PGRPS, PGRP-S, PGRP-SMGC126894, PGRPSMGC126896, TAG7, TNF superfamily, member 3 (LTB)-like (peptidoglycan recognition protein), TNFSF3L
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 200μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein PGLYRP1/PGRP-S
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.