Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse PGLYRP1/PGRP-S Protein

Catalog No. 2696PG050
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in Citrate Phosphate and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Citrate Phosphate and NaCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
2696PG050 Lyophilized from a 0.2μm filtered solution in Citrate Phosphate and NaCl with BSA as a carrier protein.
2696PG050CF Lyophilized from a 0.2μm filtered solution in Citrate Phosphate and NaCl.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 2696PG050 Supplier R&D Systems™ Supplier No. 2696PG050

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse PGLYRP1/PGRP-S Protein is derived from NS0. The Recombinant Mouse PGLYRP1/PGRP-S Protein has been validated for the following applications: Binding Activity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Citrate Phosphate and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 21946
Molecular Weight (g/mol) 19.7 kDa
Name PGRP-S
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse PGLYRP1/PGRP-S protein Cys17-Glu182,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.