Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse PlGF-2 Protein

Catalog No. 465PL050CF
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
465PL050CF 50 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
465PL010 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
465PL010CF 10 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
465PL050 50 μg Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 465PL050CF Supplier R&D Systems™ Supplier No. 465PL050/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Mouse VEGFR1/Flt-1 Fc Chimera (Catalog # 7756-FL ) is immobilized at 2 μg/mL (100 μL/well), Recombinant Mouse PlGF 2 (Catalog # 465-PL) binds with an ED50 of 0.350-3.50 ng/mL.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Gene ID (Entrez) 18654
Molecular Weight (g/mol) 15.1 kDa (monomer)
Name PlGF-2
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse PlGF-2 protein Ala24-Pro158 & Ala27-Pro158
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.