Learn More
Description
Order Info
The product is shipped at ambient temperature, upon receipt, store immediately
Specifications
Specifications
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Podocalyxin protein His22-Arg402,with a C-terminal Asp and 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
