Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Pref-1/DLK-1/FA1 Protein

Catalog No. 8545PR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8545PR050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8545PR050 Supplier R&D Systems™ Supplier No. 8545PR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Pref-1/DLK-1/FA1 Protein is derived from NS0. The Recombinant Mouse Pref-1/DLK-1/FA1 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q09163
For Use With (Application) Bioactivity
Gene ID (Entrez) 8788
Molecular Weight (g/mol) Observed M.W.: 49-66kDa, reducing conditions, Theoretical M.W.: 31kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Pref-1/DLK1/FA1 protein Ala24-Gln305,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias delta-like 1 homolog (Drosophila), DLK, DLK1, DLK-1, FA1, FA1fetal antigen 1, pG2, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG
Common Name Pref-1/DLK1/FA1
Gene Symbol Dlk1
Biological Activity Measured by its ability to inhibit adipocyte differentiation of mouse 3T3-L1 mouse embryonic fibroblast adipose-like cells. Smas, C.M. et al. (1993) Cell 73:725. Recombinant Mouse Pref-1/DLK1/FA1 at 2.5μg/mL will inhibit adipocyte differentiation by more than 50%.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Ala24-Gln305, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.