Learn More
R&D Systems™ Recombinant Mouse Protocadherin-12 Protein
Catalog No. 7926CA050
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
79-26C-A050 50 μg
1 options
This item is not returnable. View return policy
Catalog No. 79-26C-A050 Supplier R&D Systems™ Supplier No. 7926CA050
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
The Recombinant Mouse Protocadherin-12 Protein is derived from NS0. The Recombinant Mouse Protocadherin-12 Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature, upon receipt, store immediately
Specifications
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Protocadherin-12 protein Leu18-Ala716,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.