Learn More
Abcam Recombinant mouse RANKL protein (Active), 10UG
Supplier:  Abcam AB12913610UG
Encompass_Preferred
Recombinant mouse RANKL protein (Active) is a Mouse Fragment protein, in the 143 to 316 aa range, expressed in Escherichia coli, with >=95% purity, <= 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: CD254, Opgl, Rankl, Trance, Tnfsf11, Tumor necrosis factor ligand superfamily member 11, Osteoclast differentiation factor, Osteoprotegerin ligand, Receptor activator of nuclear factor kappa-B ligand, TNF-related activation-induced cytokine, ODF, OPGL, RANKL, TRANCE
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4165
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.