Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse RNF43 Protein Fc Chimera

Catalog No. 8890RN050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8890RN050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8890RN050 Supplier R&D Systems™ Supplier No. 8890RN050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse RNF43 Protein Fc Chimera is derived from NS0. The Recombinant Mouse RNF43 Protein Fc Chimera has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 54894
Molecular Weight (g/mol) Observed M.W.: 53-63kDa, reducing conditions, Theoretical M.W.: 45kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse RNF43 protein, (N-terminus) Mouse RNF43 (Ala34-Tyr197) Accession &Num; Q5NCP0 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC
Common Name RNF43
Gene Symbol Rnf43
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Mouse R-Spondin 3 is coated at 0.25μg/mL, Recombinant Mouse RNF43 Fc Chimera binds with an ED50 = 0.8-4.8ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Mouse RNF43 (Ala34-Tyr197) Accession # Q5NCP0, N-terminus; IEGRMDP; Mouse IgG2a (Glu98-Lys330), C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.