Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse ROBO2 Fc Chimera Protein

Catalog No. 8366RB050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8366RB050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 8366RB050 Supplier R&D Systems™ Supplier No. 8366RB050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse ROBO2 Fc Chimera Protein is derived from NS0. The Recombinant Mouse ROBO2 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number NP_780758
For Use With (Application) Bioactivity
Gene ID (Entrez) 6092
Molecular Weight (g/mol) Observed M.W.: 100-155kDa, reducing conditions, Theoretical M.W.: 120kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse ROBO2 protein, (N-terminus) Mouse ROBO2 (Ser22-Ala863) Accession &Num; NP_780758 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Drosophila) homolog 2, roundabout, axon guidance receptor, homolog 2 (Drosophila), SAX3
Common Name ROBO2
Gene Symbol Robo2
Biological Activity Measured by its binding ability in a functional ELISA. When recombinant human Slit3 or Recombinant Mouse Slit3 is coated at 0.25μg/mL, Recombinant Mouse ROBO2 Fc Chimera binds with an apparent Kd <2 nM.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Mouse ROBO2 (Ser22-Ala863) Accession # NP_780758, N-terminus; IEGRMDP; Mouse IgG2A (Glu98-Lys330), C-terminus
Purity or Quality Grade >90%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.