Learn More
R&D Systems™ Recombinant Mouse S100A8/S100A9 Heterodimer Protein
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | Observed M.W.: 7kDa and 12kDa, reducing conditions, Theoretical M.W.: 10kDa (S100A8), 13kDa (S100A9) |
| Quantity | 50 μg |
| Source | E. coli-derived mouse S100A8/S100A9 Heterodimer protein, (N-terminus) Mouse S100A8 (Pro2-Glu89) Accession &Num; P27005 Mouse S100A9 (Ala2-Lys113) Accession &Num; P31725 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Common Name | S100A8/S100A9 Heterodimer |
| Biological Activity | Measured by its ability to induce IL-6 secretion by A375 human melanoma cells. Hibino, T. et al. (2013) Cancer Res. 73:172. The ED50 for this effect is typically 1.5-6μg/mL. |
| Conjugate | Unconjugated |
| Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.