Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse S100A9 Protein

Catalog No. 2065S9050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
20-65S-9050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 20-65S-9050 Supplier R&D Systems™ Supplier No. 2065S9050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse S100A9 Protein is derived from E. coli. The Recombinant Mouse S100A9 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number P31725
Formulation Lyophilized from a 0.2μm filtered solution in Tris and TCEP
Gene ID (Entrez) 20202
Molecular Weight (g/mol) 13kDa
Name S100A9
Preparation Method Reconstitute at 300μg/mL in Tris and TCEP.
Quality Control Testing SDS-PAGE: 13kDa, reducing conditions
Quantity 50 μg
Source E. coli-derived mouse S100A9 protein Ala2-Lys113
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ala2
Without Additives Carrier Protein
Structural Form Noncovalently-linked homodimer
Gene Alias CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, Human S100A9 Capture, L1AG, LIAG, MAC387, MIF, MRP-14, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B)
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.