Learn More
R&D Systems™ Recombinant Mouse Semaphorin 3B Fc Chimera Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 20 with BSA as a carrier protein. |
| Gene ID (Entrez) | 20347 |
| Molecular Weight (g/mol) | 104.2 kDa (monomer) |
| Name | Semaphorin 3B Fc Chimera |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Semaphorin 3B protein, (N-terminus) Mouse Semaphorin 3B (Pro26-Asp722: Arg551Ala,Arg553Ala,Arg554Ala,Arg635Ala,Arg636Ala) Accession &Num; NP_033179 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.