Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Semaphorin 3D Fc Chimera Protein

Catalog No. 9386S3025
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μ filtered solution in Tris and NaCl with BSA as a carrier protein.
Lyophilized from a 0.2μ filtered solution in Tris and NaCl with Trehalose.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
93-86S-3025 Lyophilized from a 0.2μ filtered solution in Tris and NaCl with BSA as a carrier protein.
93-86S-3025CF Lyophilized from a 0.2μ filtered solution in Tris and NaCl with Trehalose.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 93-86S-3025 Supplier R&D Systems™ Supplier No. 9386S3025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Semaphorin 3D Fc Chimera Protein is derived from CHO. The Recombinant Mouse Semaphorin 3D Fc Chimera Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

Specifications

Accession Number Q8BH34
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μ filtered solution in Tris and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 223117
Molecular Weight (g/mol) 107 kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse Semaphorin 3D protein, (N-terminus) Mouse Semaphorin 3D (Ser38-Glu738) Accession &Num; Q8BH34 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias (semaphorin) 3D, coll-2, Collapsin 2, MGC39708
Biological Activity 1.5 to 9μg/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.