Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Citric Acid and NaCl. |
| Gene ID (Entrez) | 20350 |
| Molecular Weight (g/mol) | 111.6 kDa (monomer) |
| Name | Semaphorin 3F Fc Chimera, Truncated |
| Quantity | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Semaphorin 3F protein, (N-terminus) Mouse Semaphorin 3F Ala19-Pro775 (Arg583Ala and Arg586Ala) Accession &Num; O88632 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
