Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Semaphorin 4C Fc Chimera Protein

Catalog No. 8394S4050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83-94S-4050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 83-94S-4050 Supplier R&D Systems™ Supplier No. 8394S4050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Semaphorin 4C Fc Chimera Protein is derived from NS0. The Recombinant Mouse Semaphorin 4C Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q64151
Formulation Lyophilized from a 0.2μm filtered solution in Citric Acid and NaCl, pH 5.0
Gene ID (Entrez) 20353
Molecular Weight (g/mol) 99kDa
Name Semaphorin 4C Fc Chimera
Preparation Method Reconstitute at 200μg/mL in PBS.
Quality Control Testing SDS-PAGE: 105-125kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Semaphorin 4C protein, (N-terminus) Mouse Semaphorin 4C (Ala21-Gly664) Accession &Num; Q64151 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Ala21
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.