Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Semaphorin 4D/CD100 Fc Chimera Protein

Catalog No. 5235S4B050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-35S-4B050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 52-35S-4B050 Supplier R&D Systems™ Supplier No. 5235S4B050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

Measured by its binding ability in a functional ELISA. When Recombinant Human Plexin B2 (Catalog # 5329-PB ) is coated at 5 μg/mL, Recombinant Mouse Semaphorin 4D/CD100 Fc Chimera binds with an ED50 of 4-24 μg/mL.

Specifications

Accession Number NP_038688.2
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 10507
Molecular Weight (g/mol) M.W. (Observed): 105-135 kDa, under reducing conditions; M.W. (theoretical): 103 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse Semaphorin 4 D/CD100 protein Mouse Semaphorin 4 D/CD100 (Phe24-Met711) (N-terminus) Accession # NP_038688.2 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -70 °C under sterile conditions after opening.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Semaphorin 4 D/CD100
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.