Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Semaphorin 6A Fc Chimera Protein
SDP

Catalog No. 9017S6050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-17S-6050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-17S-6050 Supplier R&D Systems™ Supplier No. 9017S6050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Semaphorin 6A Fc Chimera Protein is derived from Sf 21 (stably transfected). The Recombinant Mouse Semaphorin 6A Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

For Use With (Application) Bioactivity
Gene ID (Entrez) 57556
Molecular Weight (g/mol) Observed M.W.: 94-120kDa, reducing conditions, Theoretical M.W.: 97kDa
Quantity 50 μg
Source Spodoptera frugiperda,Sf 21 (stably transfected)-derived mouse Semaphorin 6A protein, (N-terminus) Mouse Sema 6A (Gly19-Thr649) Accession &Num; O35464 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias KIAA1368HT018, SEMA, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6A, Sema VIA, Sema6A, SEMA6A1, SEMA6A-1, semaphorin 6A-1, Semaphorin VIA, semaphorin-6A, Semaphorin-6A-1, SEMAQ, VIA
Common Name Semaphorin 6A
Biological Activity Measured by its ability to inhibit the proliferation of HUVEC human umbilical vein endothelial cells. Moriya, J. et al. (2010) Circ. Res. 106:391. The ED50 for this effect is typically 0.2-1.2μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Spodoptera frugiperda, Sf 21 (stably transfected)-derived Mouse Sema 6A (Gly19-Thr649) Accession # O35464, N-terminus; IEGRMD; Human IgG1 (Pro100-Lys330), C-terminus
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.