Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Semaphorin 6B Fc Chimera Protein
SDP

Catalog No. 2264S6050 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-64S-6050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-64S-6050 Supplier R&D Systems™ Supplier No. 2264S6050

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Semaphorin 6B Fc Chimera Protein is derived from HEK293. The Recombinant Mouse Semaphorin 6B Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number O54951
Formulation Lyophilized from a 0.2μm filtered solution in Tris, NaCl and Tween™ 20
Gene ID (Entrez) 20359
Molecular Weight (g/mol) 91kDa (monomer)
Name Semaphorin 6B Fc Chimera
Preparation Method Reconstitute at 500μg/mL in PBS.
Quality Control Testing SDS-PAGE: 100-115kDa, reducing conditions
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived mouse Semaphorin 6B protein, (N-terminus) Mouse Semaphorin 6B (Leu27-Ser605) Accession &Num; O54951 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Leu27
Without Additives Carrier Protein
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.