Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse sFRP-1 Protein

Catalog No. 9019SF025
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-19S-F025 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 90-19S-F025 Supplier R&D Systems™ Supplier No. 9019SF025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse sFRP-1 Protein is derived from HEK293. The Recombinant Mouse sFRP-1 Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q8C4U3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 and NaCl with BSA as a carrier protein.
Gene ID (Entrez) 6422
Molecular Weight (g/mol) Observed M.W.: 37-43kDa, reducing conditions, Theoretical M.W.: 33kDa
Quantity 25 μg
Source Human embryonic kidney cell,HEK293-derived mouse sFRP-1 protein Ser32-Lys314
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias FRP1, FRP-1FrzA, FRPSARP-2, FrzA, SARP-2, SARP2sFRP-1, Secreted apoptosis-related protein 2, secreted frizzled-related protein 1, sFRP1
Common Name sFRP-1
Gene Symbol Sfrp1
Biological Activity Measured by its ability to inhibit Topflash reporter activity in HEK293T human embryonic kidney cells. The ED50 for this effect is typically 150-900ng/mL in the presence of 300ng/mL of Recombinant Mouse Wnt-3a.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Ser32-Lys314
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.