Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Siglec-G Fc Chimera Protein

Catalog No. 10103SL050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-103-SL050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-103-SL050 Supplier R&D Systems™ Supplier No. 10103SL050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Siglecs are type I transmembrane proteins that belong to the immunoglobulin (Ig) superfamily and function as mammalian lectins

  • Recombinant Protein
  • Carrier Free (CF)
  • The carrier free version does not contain Bovine Serum Albumin (BSA)

Specifications

Accession Number Q8OZE3.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 89790
Molecular Weight (g/mol) 86 kDa
Purification Method >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse Siglec-G protein Mouse Siglec-G (Met19-Lys543) (N-terminus) Accession # Q80ZE3.1 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.