Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Siglec-H Fc Chimera Protein
SDP

Catalog No. 10264SH050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-264-SH050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-264-SH050 Supplier R&D Systems™ Supplier No. 10264SH050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its binding ability in a functional ELISA.

When Recombinant Mouse Siglec-H Fc Chimera is immobilized at 2 μg/mL (100 μL/well), the concentration of Recombinant Mouse Siglec-3 Fc Chimera that produces 50% of the optimal binding response is 5 to 20 μg/mL.

Specifications

Accession Number NP_848821.2
For Use With (Application) Bioactivity
Gene ID (Entrez) 233274
Molecular Weight (g/mol) 55 kDa
Name 6430529G09Rik, SiglecH, Siglec-H, Siglech sialic acid binding Ig-like lectin H
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived mouse Siglec-H protein

Mouse Siglec-H (Asn19-Gly269) Accession # NP_848821.2IEGRMDPMouse IgG2a (Glu98-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 500 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Endotoxin Level <0.10 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.