Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Skint2/B7S3 Fc Chimera Protein

Catalog No. 10279B7050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-279-B7050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10-279-B7050 Supplier R&D Systems™ Supplier No. 10279B7050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Measured by its ability to inhibit IL-2 secretion by mouse T cells in the presence of anti-CD3. The ED50 for this effect is 0.5 to 5.0 μg/mL.

Specifications

Accession Number NP_001272892.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 329919
Molecular Weight (g/mol) 52 kDa
Quantity 50 μg
Source

Mouse myeloma cell line, NS0-derived mouse Skint2/B7S3 protein

Mouse Skint2/B7S3 (Ser22-Lys239) Accession # NP_001272892.1IEGRMDPMouse IgG2a (Glu98-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 200 μg/mL in PBS.
Buffer Lyophilized from a 0.2 μm filtered solution in PBS and NaCl.
Endotoxin Level <1.0 EU per 1 μg of the protein by the LAL method.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.