Learn More
R&D Systems™ Recombinant Mouse Slit3 (aa 1117-1523) Protein
Description
Order Info
Ships at ambient temperature
Specifications
Specifications
| Accession Number | Q9WVB4 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in MOPS and NaCl |
| Gene ID (Entrez) | 6586 |
| Molecular Weight (g/mol) | 45 kDa |
| Name | Slit3 Recombinant Protein |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Slit3 protein Leu1117-Ser1523,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.