Learn More
Description
Specifications
Specifications
| Accession Number | Q810C0 |
| For Use With (Application) | Bioactivity |
| Gene ID (Entrez) | 84631 |
| Molecular Weight (g/mol) | 69 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse SLITRK2 protein Arg22-Leu622,with a C-terminal 6-His tag |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Specific Reactivity | SLITRK2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
