Learn More
Description
Includes
For Research Use Only
Order Info
Ships at ambient temperature.
Specifications
Specifications
| Accession Number | Q70E20 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS |
| Gene ID (Entrez) | 25992 |
| Molecular Weight (g/mol) | 150 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse SNED1 protein Arg23-Lys1403,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Regulatory Status | RUO |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method |
| Biological Activity | 0.1 to 0.4μg/mL |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
