Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus

Catalog No. 464SH025CF
Change view
Click to view available options
Quantity:
200 μg
25 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT .
Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT with BSA as a carrier protein.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
464SH025CF 25 μg Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT .
464SH025 25 μg Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT with BSA as a carrier protein.
46-4SH-200 200 μg Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT with BSA as a carrier protein.
46-4SH-200CF 200 μg Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT .
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 464SH025CF Supplier R&D Systems™ Supplier No. 464SH025/CF
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus is derived from E. coli. The Recombinant Mouse Sonic Hedgehog/Shh (C25II) N-Terminus has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in NaH 2 PO 4 NaCl and DTT .
Gene ID (Entrez) 20423
Molecular Weight (g/mol) 19.8 kDa
Name Sonic Hedgehog/Shh (C25II), N-Terminus
Quantity 25 μg
Source E. coli-derived mouse Sonic Hedgehog/Shh protein Cys25-Gly198 (Cys25Ile-Ile),with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.