Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse ST3GAL5/GM3 Synthase Protein

Catalog No. 8454GT050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
84-54G-T050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 84-54G-T050 Supplier R&D Systems™ Supplier No. 8454GT050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its ability to transfer Neu5Ac from CMP-Neu5Ac to alpha-lactose. The specific activity is >5 pmol/min/μg, as measured under the described conditions.

Specifications

Accession Number O88829
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 8869
Molecular Weight (g/mol) M.W. (Observed): 45-60 kDa, reducing conditions; M.W. (theoretical): 38 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse ST3GAL5/GM3 Synthase protein Thr87-His414, with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Endotoxin Concentration <0.1 EU / 1 μg of the protein by the LAL method.
Gene Alias 3S-T, alpha 2,3-sialyltransferase V, CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase, EC 2.4.99.9, Ganglioside GM3 synthase, GM3 synthase, lactosylceramide alpha-2,3-sialyltransferase, SATI, Sialyltransferase 9, sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase;GM3 synthase), Siat9, SIATGM3S, ST3 beta-galactoside alpha-2,3-sialyltransferase 5, ST3Gal V, ST3GALV, ST3GalVSIAT9SATI
Common Name ST3GAL5
Gene Symbol St3gal5
Biological Activity Measured by its ability to transfer Neu5Ac from CMP-Neu5Ac to alpha-lactose. The specific activity is >5 pmol/min/μg, as measured under the described conditions.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Thr87-His414, with an N-terminal 6-His tag
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain (at 5 μg / lane)
Protein ST3GAL5
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.