Learn More
R&D Systems™ Recombinant Mouse TACE/ADAM17 Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in MES and NaCl. |
| Gene ID (Entrez) | 11491 |
| Molecular Weight (g/mol) | 63 kDa, 40 kDa, 23 kDa, 18 kDa |
| Name | TACE/ADAM17 |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse TACE/ADAM17 protein Pro18-Asp563 and Arg215-Asp563,both with a C-terminal 10-His tag,Pro18-Arg214 and Asp59-Arg214 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Recombinant Mouse TACE/ADAM17 is prone to proteolytic cleavage at C-terminus. The poly-His tag may not be present in the preparation. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.