Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse TGF-beta 1 Protein
GSA_VASDP

Catalog No. 7666MB005CF Shop All Novus Biologicals Products
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in HCl.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation
7666MB005CF Lyophilized from a 0.2μm filtered solution in HCl.
7666MB005 Lyophilized from a 0.2μm filtered solution in HCl with BSA as a carrier protein.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 7666MB005CF Supplier R&D Systems™ Supplier No. 7666MB005/CF

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse TGF-beta 1 Protein is derived from CHO. The Recombinant Mouse TGF-beta 1 Protein has been validated for the following applications: Bioactivity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in HCl.
Gene ID (Entrez) 21803
Molecular Weight (g/mol) 12.8 kDa (monomer)
Name TGF-beta 1
Quantity 5 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse TGF-beta 1 protein Ala279-Ser390
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.