Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse Thrombopoietin Protein

Catalog No. 488TO200
Change view
Click to view available options
Quantity:
200 μg
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Formulation
48-8TO-200 200 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
488TO005 5 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
488TO005CF 5 μg Lyophilized from a 0.2μm filtered solution in PBS.
488TO025 25 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
488TO025CF 25 μg Lyophilized from a 0.2μm filtered solution in PBS.
48-8TO-200CF 200 μg Lyophilized from a 0.2μm filtered solution in PBS.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 48-8TO-200 Supplier R&D Systems™ Supplier No. 488TO200
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse Thrombopoietin Protein is derived from NS0. The Recombinant Mouse Thrombopoietin Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 21832
Molecular Weight (g/mol) 35 kDa
Name Thrombopoietin/Tpo
Quantity 200 μg
Source Mouse myeloma cell line,NS0-derived mouse Thrombopoietin/Tpo protein Ser22-Thr356
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.