Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse TIGIT Fc Chimera Protein

Catalog No. 7267TG01M
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-67T-G01M 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 72-67T-G01M Supplier R&D Systems™ Supplier No. 7267TG01M
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Measured by its binding ability in a functional ELISA. When Recombinant Mouse CD155/PVR (Catalog # 6909-CD ) is coated at 2.5 μg/mL (100 μL/well), the concentration of Recombinant Mouse TIGIT Fc Chimera that produces 50% optimal binding response is 10-50 ng/mL.

Specifications

Accession Number NP_001139797
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 201633
Molecular Weight (g/mol) M.W. (Observed): 50-60 kDa, reducing conditions; M.W. (theoretical): 40.1 kDa (monomer)
Quantity 1 mg
Source Mouse myeloma cell line, NS0-derived mouse TIGIT protein Mouse TIGIT (Gly26-Thr143) (N-terminus) Accession # NP_001139797 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Symbol Tigit
Product Type Recombinant Protein
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 100 μg/mL in PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein TIGIT
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.