Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse TIM-1/KIM-1/HAVCR Isoform A Fc Protein
SDP

Catalog No. 10699TM050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10699TM050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 10699TM050 Supplier R&D Systems™ Supplier No. 10699TM050
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Measured in a cell proliferation assay using PHA-stimulated human T lymphoblasts. The ED50 for this effect is 0.5-4.0 μg/mL.. Measured by its binding ability in a functional ELISA. When Recombinant Mouse TIM-1/KIM-1/HAVCR Isoform A Fc Chimera (Catalog # 10699-TM) is immobilized at 1 μg/mL (100 μL/well), Recombinant Mouse TIM-4 (Catalog # 2826-TI ) binds with an ED50 of 0.8-6.4 μg/mL.

Specifications

Accession Number NP_599009.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 26762
Molecular Weight (g/mol) M.W.-observed: 75-85 kDa, under reducing conditions, M.W.-theroretical: 50 kDa
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse TIM-1/KIM-1/HAVCR protein Mouse TIM-1/KIM-1/HAVCR (Tyr22-Gly237) (N-terminus) Accession # NP_599009.2 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1 TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein TIM-1/KIM-1/HAVCR
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.