Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse TINAGL1 Protein

Catalog No. 9208TI050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-08T-I050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 92-08T-I050 Supplier R&D Systems™ Supplier No. 9208TI050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse TINAGL1 Protein is derived from HEK293. The Recombinant Mouse TINAGL1 Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

Specifications

Accession Number Q99JR5
For Use With (Application) Bioactivity
Formulation Supplied as a 0.2μm filtered solution in Tris and NaCl.
Gene ID (Entrez) 64129
Molecular Weight (g/mol) 49 kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived mouse TINAGL1 protein Asp41-His466,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Biological Activity Measured by its binding ability in a functional ELISA. When Cultrex Mouse Laminin Iis coated at 5 μg/mL, Recombinant Mouse TINAGL1 binds with an ED50 = 3-18 ng/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.