Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse TLR4 Fc Chimera Protein

Catalog No. 9149TR050
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-49T-R050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 91-49T-R050 Supplier R&D Systems™ Supplier No. 9149TR050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse TLR4 Fc Chimera Protein is derived from CHO. The Recombinant Mouse TLR4 Fc Chimera Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q9QUK6
For Use With (Application) Bioactivity
Gene ID (Entrez) 7099
Molecular Weight (g/mol) Observed M.W.: 115-137kDa, reducing conditions, Theoretical M.W.: 96kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse TLR4 protein, (N-terminus) Mouse TLR4 (Asn26-Lys629) Accession &Num; Q9QUK6 DIEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias ARMD10, CD284, CD284 antigen, hTollhomolog of Drosophila toll, TOLL, toll-like receptor 4
Common Name TLR4
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Mouse TLR4 Fc Chimera is immobilized at 2μg/mL, 100μL/well, the concentration of Recombinant Human MD-2 that produces 50% of the optimal binding response is approximately 0.15-0.9μg/mL
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Mouse TLR4 (Asn26-Lys629) Accession # Q9QUK6, N-terminus; DIEGRMDP; Mouse IgG2a (Glu98-Lys330), C-terminus
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.