Learn More
R&D Systems™ Recombinant Mouse TMEFF2/Tomoregulin-2 Fc Protein
Description
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 23671 |
| Molecular Weight (g/mol) | M.W.-observed: 77-90 kDa, under reducing conditions., M.W.-theroretical: 58 kDa |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line, NS0-derived mouse TMEFF2/Tomoregulin-2 protein Mouse TMEFF2 (Phe41-Val320) (C-terminus) Accession # Q9QYM9.1 IEGRMDP Mouse IgG2A (Glu98-Lys330) (N-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2 |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.