Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse TRAIL/TNFSF10 Protein

Catalog No. 1121TL010
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in Tris, Ammonium Sulfate and ZnSO 4 with Trehalose and with BSA as a carrier protein.
Supplied as a 0.2 μm filtered solution in Tris, Ammonium Sulfate and ZnSO 4 .
Molecular Weight (g/mol):
20 kDa (monomer)
20 kda
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Formulation Molecular Weight (g/mol)
1121TL010 Lyophilized from a 0.2 μm filtered solution in Tris, Ammonium Sulfate and ZnSO 4 with Trehalose and with BSA as a carrier protein. 20 kda
1121TL010CF Supplied as a 0.2 μm filtered solution in Tris, Ammonium Sulfate and ZnSO 4 . 20 kDa (monomer)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 1121TL010 Supplier R&D Systems™ Supplier No. 1121TL010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse TRAIL/TNFSF10 Protein is derived from E. coli. The Recombinant Mouse TRAIL/TNFSF10 Protein has been validated for the following applications: Bioactivity.

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in Tris, Ammonium Sulfate and ZnSO 4 with Trehalose and with BSA as a carrier protein.
Gene ID (Entrez) 22035
Molecular Weight (g/mol) 20 kda
Name TRAIL/TNFSF10
Quantity 10 μg
Source E. coli-derived mouse TRAIL/TNFSF10 protein Pro118-Asn291
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.