Learn More
R&D Systems™ Recombinant Mouse TRANCE/RANK L/TNFSF11 Protein
Description
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 21943 |
| Molecular Weight (g/mol) | 28 kDa |
| Name | TRANCE/TNFSF11/RANK L |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse TRANCE/TNFSF11/RANK L protein Arg72-Asp316,with an N-terminal 6-His tag |
| Storage Requirements | 3 months from date of receipt, 2 to 8° C as supplied. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.