Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Mouse TREM2 His-tag Protein

Catalog No. 9228T2050
Change view
Click to view available options
Recombinant Mouse TREM-2 His-tag Protein, Carrier-Free
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
92-28T-2050 Recombinant Mouse TREM-2 His-tag Protein, Carrier-Free
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 92-28T-2050 Supplier R&D Systems™ Supplier No. 9228T2050
Add to Cart
Add to Cart
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse TREM-2 His-tag Protein is derived from NS0. The Recombinant Mouse TREM-2 His-tag Protein has been validated for the following applications: Bioactivity.

Order Info

The product is shipped at ambient temperature.

Specifications

Accession Number Q99NH8
For Use With (Application) Bioactivity
Molecular Weight (g/mol) M.W. (Observed): 25 - 75 kDa, under reducing conditions.
Name TREM2
Source Mouse myeloma cell line, NS0-derived mouse TREM2 protein Leu19-Ser171, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of protein by LAL method
Gene Alias PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2
Biological Activity Measured by its ability to bind fluorescein-conjugated S. aureus Bioparticles. Daws, M.R.(2003) J. Immunol.The ED50 for this effect is 20-120 ng/mL using a His tag antibody coated plate.
Conjugate Unconjugated

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.