Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse TROP-2 His (C-Term) Protein
SDP

Catalog No. NB171831 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB171831 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB171831 Supplier Novus Biologicals™ Supplier No. NBP318209
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Specifications

Concentration LYOPH
For Use With (Application) SDS-PAGE
Formulation Lyophilized from a 0.22 um filtered solution of PBS, pH 7.4
Gene ID (Entrez) 4070
Quantity 100 μg
Storage Requirements Store at -20°C.
Endotoxin Concentration <0.1 EU/μg
Gene Alias Cell surface glycoprotein Trop-2, EGP1, EGP-1, epithelial glycoprotein-1, GA733-1GP50, gastrointestinal tumor-associated antigen GA7331, M1S1cell surface glycoprotein TROP2, Membrane component chromosome 1 surface marker 1, membrane component, chromosome 1, surface marker 1, Pancreatic carcinoma marker protein GA733-1, pancreatic carcinoma marker protein GA7331,40kD glycoprotein, identified by monoclonal GA733, TROP2GA7331, tumor-associated calcium signal transducer 2
Gene Symbol TACSTD2
Reconstitution Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water
Research Category Signal Transduction
Purity or Quality Grade >95%, by SDS-PAGE
Protein TROP-2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.